Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abl1 | NM_001112703.2 | NP_001106174.1 | ||
| NM_001283045.1 | NP_001269974.1 | |||
| NM_001283046.1 | NP_001269975.1 | |||
| NM_001283047.1 | NP_001269976.1 | |||
| NM_009594.4 | NP_033724.2 | |||
| AK088252.1 | BAE43394.1 | |||
| AK090095.1 | BAC41088.1 | |||
| AK166028.1 | ||||
| AK166210.1 | BAE38629.1 | |||
| AK190971.1 | ||||
| AK218428.1 | ||||
| BC059260.1 | AAH59260.1 | |||
| BC103770.1 | AAI03771.1 | |||
| CF727094.1 | ||||
| CK626997.1 | ||||
| J02995.1 | AAA88241.1 | |||
| L10656.1 |

Genomic Map