Assay Details
Target Gene Details
Entrez Gene ID: | 108897 |
Gene Name: | allograft inflammatory factor 1-like |
Gene Aliases: |
2810003C17Rik, AI043124, C87647, Iba2 |
Location: |
Chr.2:31950303-31973442 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aif1l | NM_145144.1 | 5 | 329 | NP_660126.1 |
| AB035322.1 | 5 | 281 | BAB20749.1 | |
| AK028955.1 | 5 | 287 | BAC26211.1 | |
| AK045539.1 | 5 | 359 | BAC32410.1 | |
| BC024599.1 | 5 | 329 | AAH24599.1 |
More Information
Set Membership: |
|

Genomic Map