Assay Details
Target Gene Details
Entrez Gene ID: | 67126 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, epsilon subunit |
Gene Aliases: |
2410043G19Rik, ATPE, AV000645 |
Location: |
Chr.2:174461075-174464101 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5e | NM_025983.3 | NP_080259.1 | ||
| AK007752.1 | ||||
| AK007809.1 | BAB25274.1 | |||
| AK010661.2 | BAB27098.1 | |||
| AK012444.1 | BAB28243.1 | |||
| AK160387.1 | BAE35759.1 | |||
| AY341882.1 | AAQ24166.1 | |||
| BC024339.1 | AAH24339.1 | |||
| CJ322223.1 | ||||
| W78532.1 |
More Information
Set Membership: |
|

Genomic Map