Assay Details
Target Gene Details
Entrez Gene ID: | 228033 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F0 complex, subunit C3 (subunit 9) |
Gene Aliases: |
6030447M23 |
Location: |
Chr.2:73908447-73911326 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp5g3 | NM_001301721.1 | NP_001288650.1 | ||
| NM_001301722.1 | NP_001288651.1 | |||
| NM_175015.3 | NP_778180.1 | |||
| AK077930.1 | BAC37071.1 | |||
| AK087955.1 | BAC40056.1 | |||
| AW554443.2 | ||||
| BC116218.2 | ||||
| CF104340.1 | ||||
| CX208968.1 |
More Information
Set Membership: |
|

Genomic Map