Assay Details
Target Gene Details
Entrez Gene ID: | 109778 |
Gene Name: | biliverdin reductase A |
Gene Aliases: |
0610006A11Rik, 2500001N03Rik, Blvr |
Location: |
Chr.2:127070629-127097084 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blvra | NM_026678.4 | NP_080954.4 | ||
| AK010847.1 | BAB27219.1 | |||
| AK045022.1 | ||||
| AK162186.1 | BAE36777.1 | |||
| AK172620.1 | BAE43098.1 | |||
| BC052146.1 | AAH52146.1 | |||
| BY073963.1 |
More Information
Set Membership: |
|

Genomic Map