Assay Details
Target Gene Details
Entrez Gene ID: | 229003 |
Gene Name: | helicase with zinc finger 2, transcriptional coactivator |
Gene Aliases: |
PDIP1, Pric285, mKIAA1769, mPDIP1 |
Location: |
Chr.2:181227615-181242058 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Helz2 | NM_183162.2 | NP_898985.2 | ||
| AB201714.1 | ||||
| AK173259.1 | BAD32537.1 | |||
| BC006779.1 | AAH06779.1 |
More Information
Set Membership: |
|

Genomic Map