Assay Details
Target Gene Details
Entrez Gene ID: | 320664 |
Gene Name: | Cas scaffolding protein family member 4 |
Gene Aliases: |
F730031O20Rik |
Location: |
Chr.2:172393683-172433757 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cass4 | NM_001080820.2 | NP_001074289.1 | ||
| NM_001276422.1 | NP_001263351.1 | |||
| NM_001276423.1 | NP_001263352.1 | |||
| AK036419.1 | ||||
| AK080960.1 | ||||
| AK089450.1 | ||||
| AK170726.1 | BAE41983.1 | |||
| BC117858.1 | ||||
| BC117859.1 | ||||
| BC129975.1 | ||||
| BC129976.1 |

Genomic Map