Assay Details
Target Gene Details
Entrez Gene ID: | 104010 |
Gene Name: | cadherin 22 |
Gene Aliases: |
- |
Location: |
Chr.2:165111501-165234879 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh22 | NM_174988.3 | 6 | 1460 | NP_778153.2 |
| AB019618.1 | 6 | 1450 | BAA34426.1 | |
| AK140248.1 | 6 | 1578 |
More Information
Set Membership: |
|

Genomic Map