Assay Details
Target Gene Details
Entrez Gene ID: | 107951 |
Gene Name: | cyclin-dependent kinase 9 (CDC2-related kinase) |
Gene Aliases: |
PITALRE |
Location: |
Chr.2:32705782-32712784 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk9 | NM_130860.3 | NP_570930.1 | ||
| AF327431.1 | AAK15699.1 | |||
| AK089276.1 | BAC40824.1 | |||
| AK142397.1 | BAE25055.1 | |||
| AK143217.1 | BAE25312.1 | |||
| AK144607.1 | BAE25966.1 | |||
| AK155733.1 | ||||
| AK157340.1 | BAE34054.1 | |||
| BC003901.1 | AAH03901.1 |
More Information
Set Membership: |
|

Genomic Map