Assay Details
Target Gene Details
Entrez Gene ID: | 99151 |
Gene Name: | cerebral endothelial cell adhesion molecule |
Gene Aliases: |
2600006K01Rik, AL024097, Ceecam1 |
Location: |
Chr.2:29845105-29882845 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cercam | NM_207298.2 | NP_997181.1 | ||
| AK155384.1 | ||||
| BC138847.1 |
More Information
Set Membership: |
|

Genomic Map