Assay Details
Target Gene Details
Entrez Gene ID: | 68379 |
Gene Name: | CDKN1A interacting zinc finger protein 1 |
Gene Aliases: |
0610038H21Rik, 2900056O04Rik, LSFR1, ZNF356 |
Location: |
Chr.2:32363006-32378313 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciz1 | NM_001252534.1 | 11 | 1922 | NP_001239463.1 |
| NM_001252536.1 | 12 | 1634 | NP_001239465.1 | |
| NM_001252537.1 | 11 | 1895 | NP_001239466.1 | |
| NM_001252538.1 | 11 | 1585 | NP_001239467.1 | |
| NM_028412.2 | 10 | 1699 | NP_082688.1 | |
| AJ575057.1 | 9 | 1206 | CAE00633.1 | |
| AK089986.1 | 11 | 1898 | BAC41030.1 | |
| AK137961.1 | 11 | 1501 | BAE23524.1 | |
| AK159103.1 | 11 | 1900 | BAE34819.1 | |
| AK165687.1 | 12 | 1588 | BAE38339.1 | |
| BC018483.1 | 10 | 1663 | AAH18483.1 | |
| CX217150.1 | 3 | 410 |

Genomic Map