Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dab2ip | NM_001114125.1 | NP_001107597.1 | ||
| DQ473307.1 |
Target Gene Details
Entrez Gene ID: | 100042194 |
Gene Name: | predicted gene 13446 |
Gene Aliases: |
OTTMUSG00000012261 |
Location: |
Chr.2:35549534-35558702 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm13446 | NR_045894.1 | 1 | 87 | |
| AK138201.1 | 1 | 88 |
More Information
Set Membership: |
|

Genomic Map