Assay Details
Target Gene Details
Entrez Gene ID: | 69601 |
Gene Name: | disabled 2 interacting protein |
Gene Aliases: |
2310011D08Rik, AI480459, Aip1, mKIAA1743 |
Location: |
Chr.2:35558458-35730994 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dab2ip | NM_001001602.2 | 7 | 1595 | NP_001001602.2 |
| NM_001114124.2 | 9 | 1719 | NP_001107596.1 | |
| NM_001114125.1 | 9 | 1571 | NP_001107597.1 | |
| NM_001290637.1 | 9 | 1626 | NP_001277566.1 | |
| NM_001290639.1 | 7 | 1254 | NP_001277568.1 | |
| NM_001290640.1 | 7 | 1559 | NP_001277569.1 | |
| NM_001290641.1 | 9 | 1852 | NP_001277570.1 | |
| AK122548.1 | 4 | 559 | BAC65830.1 | |
| AK147464.1 | 9 | 1651 | BAE27930.1 | |
| AK147593.1 | 9 | 1853 | BAE28013.1 | |
| AK155296.1 | 9 | 1627 | BAE33173.1 | |
| AY305656.1 | 7 | 1312 | AAQ77379.1 | |
| AY305657.1 | 7 | 1595 | AAQ77380.1 | |
| AY305658.1 | 7 | 1254 | AAQ77381.1 | |
| BC094373.1 | 4 | 416 | AAH94373.1 | |
| BC118530.1 | 7 | 1220 | ||
| DQ473307.1 | 9 | 1456 |
More Information
Set Membership: |
|

Genomic Map