Assay Details
Target Gene Details
Entrez Gene ID: | 66044 |
Gene Name: | D-tyrosyl-tRNA deacylase 1 |
Gene Aliases: |
0610006H08Rik, DUE-B, Hars2 |
Location: |
Chr.2:144599953-144768747 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtd1 | NM_025314.3 | 1 | 109 | NP_079590.1 |
| AK002246.1 | ||||
| AK003594.1 | 1 | 81 | BAB22882.1 | |
| AK010822.1 | BAB27205.1 | |||
| AK011917.1 | 1 | 77 | BAB27914.1 | |
| AK017565.1 | BAB30808.1 | |||
| AK158898.1 | 1 | 166 | BAE34719.1 | |
| BC026537.1 | 1 | 82 | AAH26537.1 | |
| BY294360.1 | 1 | 109 |
More Information
Set Membership: |
|

Genomic Map