Assay Details
Target Gene Details
Entrez Gene ID: | 13480 |
Gene Name: | dolichol-phosphate (beta-D) mannosyltransferase 1 |
Gene Aliases: |
AI118379, AI194292 |
Location: |
Chr.2:168209048-168230379 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpm1 | NM_010072.3 | 1 | 136 | NP_034202.1 |
| AB004789.1 | 1 | 110 | BAA25759.1 | |
| AI099209.1 | 1 | 136 | ||
| AK004834.1 | 1 | 129 | BAB23602.1 | |
| AK005268.1 | 1 | 122 | BAB23920.1 | |
| AK008548.1 | 1 | 124 | BAB25735.1 | |
| BC061151.1 | 1 | 116 | AAH61151.1 |
More Information
Set Membership: |
|

Genomic Map