Assay Details
Target Gene Details
Entrez Gene ID: | 13480 |
Gene Name: | dolichol-phosphate (beta-D) mannosyltransferase 1 |
Gene Aliases: |
AI118379, AI194292 |
Location: |
Chr.2:168209048-168230379 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpm1 | NM_010072.3 | NP_034202.1 | ||
| AB004789.1 | BAA25759.1 | |||
| AI099209.1 | ||||
| AK004834.1 | BAB23602.1 | |||
| AK005268.1 | BAB23920.1 | |||
| AK008548.1 | BAB25735.1 | |||
| AK147481.1 | 1 | 951 | ||
| BC061151.1 | AAH61151.1 |
More Information
Set Membership: |
|

Genomic Map