Assay Details
Target Gene Details
Entrez Gene ID: | 228836 |
Gene Name: | discs, large homolog-associated protein 4 (Drosophila) |
Gene Aliases: |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
Location: |
Chr.2:156613633-156764363 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap4 | NM_001277186.1 | 7 | 2027 | NP_001264115.1 |
| NM_001277187.1 | 6 | 1976 | NP_001264116.1 | |
| NM_146128.6 | 7 | 2027 | NP_666240.4 | |
| AK122408.1 | 7 | 2027 | BAC65690.2 | |
| AY243849.2 | 5 | 1607 | AAO89220.2 | |
| BC141110.1 | 5 | 1664 | ||
| BC145394.1 | 5 | 1664 | ||
| BC145395.1 | 4 | 1613 |
More Information
Set Membership: |
|

Genomic Map