Assay Details
Target Gene Details
Entrez Gene ID: | 13804 |
Gene Name: | endonuclease G |
Gene Aliases: |
- |
Location: |
Chr.2:30171401-30174069 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Endog | NM_007931.1 | NP_031957.1 | ||
| AB012108.1 | BAA28168.1 | |||
| AK144459.1 | BAE25900.1 | |||
| BC030177.1 | AAH30177.1 | |||
| X99394.1 | CAA67768.1 |

Genomic Map