Assay Details
Target Gene Details
Entrez Gene ID: | 252864 |
Gene Name: | dual specificity phosphatase-like 15 |
Gene Aliases: |
AI851682, LMW-DSP10, T-DSP10 |
Location: |
Chr.2:152940995-152952277 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dusp15 | NM_001159376.1 | NP_001152848.1 | ||
| NM_145744.2 | NP_665687.1 | |||
| AF357887.1 | AAM00226.1 | |||
| BC116841.1 | ||||
| BC116843.1 | ||||
| BC144711.1 |
More Information
Set Membership: |
|

Genomic Map