Assay Details
Target Gene Details
Entrez Gene ID: | 228592 |
Gene Name: | RIKEN cDNA F830045P16 gene |
Gene Aliases: |
Sirpb3 |
Location: |
Chr.2:129457797-129536602 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F830045P16Rik | NM_177653.3 | NP_808321.1 | ||
| AK089892.1 | BAC40988.1 |
More Information
Set Membership: |
|

Genomic Map