Assay Details
Target Gene Details
Entrez Gene ID: | 319513 |
Gene Name: | PC-esterase domain containing 1A |
Gene Aliases: |
A930025D01Rik, AI480484, Fam113a |
Location: |
Chr.2:130417684-130424641 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pced1a | NM_001114541.1 | NP_001108013.1 | ||
| AK031469.1 | 1 | 100 | BAC27419.1 | |
| BC064802.1 | 1 | 85 | AAH64802.1 | |
| BY326318.1 |
Target Gene Details
Entrez Gene ID: | 80743 |
Gene Name: | vacuolar protein sorting 16 (yeast) |
Gene Aliases: |
1810074M16Rik |
Location: |
Chr.2:130424304-130444269 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vps16 | NM_030559.3 | NP_085036.3 | ||
| AB056721.1 | BAB64892.1 | |||
| AK049872.1 | BAC33966.1 | |||
| AK162984.1 | ||||
| AK169694.1 | ||||
| BC025626.1 | AAH25626.1 | |||
| BY123989.1 |
More Information
Set Membership: |
|

Genomic Map