Assay Details
Target Gene Details
Entrez Gene ID: | 74562 |
Gene Name: | fer-1-like 4 (C. elegans) |
Gene Aliases: |
9130402C12Rik |
Location: |
Chr.2:156019139-156053018 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 39 - Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fer1l4 | NM_001136556.1 | NP_001130028.1 | ||
| AK018656.1 | BAB31328.1 | |||
| AK035669.1 |
More Information
Set Membership: |
|

Genomic Map