Assay Details
Target Gene Details
Entrez Gene ID: | 433485 |
Gene Name: | synapse differentiation inducing 1 |
Gene Aliases: |
AW123113, DSPC2, Gm14134, H2C20orf39, OTTMUSG00000015750, Tmem90b |
Location: |
Chr.2:149830053-150004392 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syndig1 | NM_001085521.2 | NP_001078990.1 | ||
| BB653505.1 | ||||
| CJ061528.1 |
More Information
Set Membership: |
|

Genomic Map