Assay Details
Target Gene Details
Entrez Gene ID: | 433485 |
Gene Name: | synapse differentiation inducing 1 |
Gene Aliases: |
AW123113, DSPC2, Gm14134, H2C20orf39, OTTMUSG00000015750, Tmem90b |
Location: |
Chr.2:149830053-150004392 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syndig1 | NM_001085521.2 | NP_001078990.1 | ||
| BC147352.1 | ||||
| BC147353.1 | ||||
| BC147660.1 | ||||
| BC147661.1 |

Genomic Map