Assay Details
Target Gene Details
Entrez Gene ID: | 271786 |
Gene Name: | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 |
Gene Aliases: |
A230002A12, A230020F20, BB182356 |
Location: |
Chr.2:54436387-55118309 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Galnt13 | NM_173030.2 | 6 | 1092 | NP_766618.2 |
| AB082928.1 | 4 | 580 | BAC54546.1 | |
| AK030726.1 | 5 | 1023 | ||
| AK038387.1 | 6 | 1093 | BAC29981.1 | |
| AK045417.1 | 6 | 1093 | BAC32353.1 | |
| BC131651.1 | 6 | 771 | ||
| BC131652.1 | 5 | 802 |
More Information
Set Membership: |
|

Genomic Map