Assay Details
Target Gene Details
Entrez Gene ID: | 70933 |
Gene Name: | RIKEN cDNA 4931402G19 gene |
Gene Aliases: |
- |
Location: |
Chr.2:120455698-120469367 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4931402G19Rik | NR_040608.1 | 5 | 1945 | |
| AK016423.1 | 5 | 1946 |
Target Gene Details
Entrez Gene ID: | 76051 |
Gene Name: | glucosidase, alpha; neutral C |
Gene Aliases: |
5830445O15Rik, 9330160A12, mFLJ00088 |
Location: |
Chr.2:120403896-120460866 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ganc | NM_172672.2 | NP_766260.2 | ||
| AK017994.1 | ||||
| AK076333.1 | BAC36303.1 | |||
| AK220303.1 | BAD90228.1 |
More Information
Set Membership: |
|

Genomic Map