Assay Details
Target Gene Details
Entrez Gene ID: | 228911 |
Gene Name: | teashirt zinc finger family member 2 |
Gene Aliases: |
2900073F20Rik, B830045G17, Mtsh2, Sdccag33l, Tsh2, Zfp218, mKIAA4248, teashirt2 |
Location: |
Chr.2:169632496-170071666 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tshz2 | AK045693.1 | BAC32459.1 | ||
| AK046877.1 | ||||
| AK161999.1 | BAE36673.1 |

Genomic Map