Assay Details
Target Gene Details
Entrez Gene ID: | 15139 |
Gene Name: | hemolytic complement |
Gene Aliases: |
C5, C5a, He |
Location: |
Chr.2:34983331-35061441 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hc | NM_010406.2 | 17 | 2279 | NP_034536.1 |
| AK080166.1 | BAC37836.1 | |||
| M35525.1 | 17 | 2234 | AAA37349.1 | |
| M35526.1 | 17 | 2232 | AAA37348.1 |
More Information
Set Membership: |
|

Genomic Map