Assay Details
Target Gene Details
Entrez Gene ID: | 67839 |
Gene Name: | G-protein signalling modulator 1 (AGS3-like, C. elegans) |
Gene Aliases: |
1810037C22Rik, AW107933, Ags3 |
Location: |
Chr.2:26314606-26348237 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gpsm1 | NM_001199146.1 | 6 | 867 | NP_001186075.1 |
| NM_001199147.1 | 6 | 936 | NP_001186076.1 | |
| NM_153410.5 | 6 | 867 | NP_700459.2 | |
| AK048691.1 | 6 | 861 | BAC33422.1 | |
| AK082985.1 | 6 | 810 | ||
| AK084631.1 | 6 | 809 | BAC39236.1 | |
| BC026486.1 | AAH26486.1 | |||
| BC027004.1 | 4 | 374 | AAH27004.1 | |
| BC071197.1 | 6 | 933 | AAH71197.1 |

Genomic Map