Assay Details
Target Gene Details
| Entrez Gene ID: | 67839 | 
| Gene Name: | G-protein signalling modulator 1 (AGS3-like, C. elegans) | 
| Gene Aliases: | 1810037C22Rik, AW107933, Ags3 | 
| Location: | Chr.2:26314606-26348237 on Build GRCm38 | 
| Assay Gene Location: | Overlaps Intron 8 - Exon 9 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| Gpsm1 | NM_001199146.1 | NP_001186075.1 | ||
| NM_001199147.1 | NP_001186076.1 | |||
| NM_153410.5 | NP_700459.2 | |||
| AK048691.1 | BAC33422.1 | |||
| AK082985.1 | ||||
| AK084631.1 | BAC39236.1 | |||
| AK192364.1 | ||||
| BC026486.1 | AAH26486.1 | |||
| BC027004.1 | AAH27004.1 | |||
| BC071197.1 | AAH71197.1 | 
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top Intragenic
							Intragenic