Assay Details
Target Gene Details
Entrez Gene ID: | 15162 |
Gene Name: | hemopoietic cell kinase |
Gene Aliases: |
AI849071, Bmk |
Location: |
Chr.2:153108468-153151441 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hck | NM_001172117.1 | NP_001165588.1 | ||
| NM_010407.4 | NP_034537.2 | |||
| AK149736.1 | BAE29054.1 | |||
| AK150290.1 | BAE29445.1 | |||
| AK150709.1 | BAE29787.1 | |||
| AK152506.1 | ||||
| AK155975.1 | BAE33532.1 | |||
| AK159234.1 | ||||
| AK165315.1 | BAE38133.1 | |||
| BC010478.1 | ||||
| J03023.1 | AAA37305.1 | |||
| Y00487.1 | CAA68544.1 |

Genomic Map