Assay Details
Target Gene Details
Entrez Gene ID: | 227753 |
Gene Name: | gelsolin |
Gene Aliases: |
ADF |
Location: |
Chr.2:35256359-35307902 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gsn | NM_001206367.1 | 13 | 1714 | NP_001193296.1 |
| NM_001206368.1 | 12 | 1630 | NP_001193297.1 | |
| NM_001206369.1 | 12 | 1675 | NP_001193298.1 | |
| NM_146120.4 | 12 | 1737 | NP_666232.2 | |
| AK076156.1 | 12 | 1677 | BAC36223.1 | |
| AK089934.1 | 13 | 1669 | BAC41004.1 | |
| AK135632.1 | 12 | 1450 | ||
| AK143664.1 | 12 | 1704 | BAE25485.1 | |
| AK146969.1 | 13 | 1631 | BAE27574.1 | |
| AK148957.1 | 13 | 1694 | BAE28702.1 | |
| AK150359.1 | 13 | 1637 | BAE29494.1 | |
| AK151684.1 | 13 | 1656 | BAE30608.1 | |
| AK153029.1 | 13 | 1634 | BAE31661.1 | |
| AK166831.1 | 12 | 1650 | BAE39054.1 | |
| AK166932.1 | 13 | 1628 | BAE39126.1 | |
| AK167918.1 | 13 | 1644 | BAE39925.1 | |
| AK168461.1 | 13 | 1647 | BAE40358.1 | |
| AK168532.1 | 13 | 1648 | BAE40410.1 | |
| AK168707.1 | 12 | 1583 | BAE40549.1 | |
| AK168725.1 | 13 | 1631 | BAE40566.1 | |
| AK168829.1 | 12 | 1682 | ||
| AK169041.1 | 13 | 1634 | BAE40830.1 | |
| AK169670.1 | 13 | 1659 | BAE41291.1 | |
| AK188287.1 | ||||
| AK188608.1 | ||||
| AK212201.1 | ||||
| BC023143.1 | 12 | 1675 | ||
| BC060377.1 | 13 | 1662 | AAH60377.1 | |
| J04953.1 | 11 | 1524 | AAA37677.1 |
More Information
Set Membership: |
|

Genomic Map