Assay Details
Target Gene Details
Entrez Gene ID: | 16403 |
Gene Name: | integrin alpha 6 |
Gene Aliases: |
5033401O05Rik, AI115430, Cd49f, VLA-6 |
Location: |
Chr.2:71786885-71858427 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itga6 | NM_001277970.1 | NP_001264899.1 | ||
| NM_008397.4 | NP_032423.2 | |||
| AK034186.1 | BAC28623.1 | |||
| AK045391.1 | BAC32340.1 | |||
| AK083717.1 | BAC39003.1 | |||
| AK159653.1 | BAE35264.1 | |||
| BC049132.1 | AAH49132.1 | |||
| BC058095.1 | AAH58095.1 | |||
| CO430533.1 | ||||
| X69902.1 | CAA49527.1 |
More Information
Set Membership: |
|

Genomic Map