Assay Details
Target Gene Details
Entrez Gene ID: | 16410 |
Gene Name: | integrin alpha V |
Gene Aliases: |
1110004F14Rik, 2610028E01Rik, CD51, D430040G12Rik |
Location: |
Chr.2:83723819-83806917 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgav | NM_008402.3 | 17 | 1982 | NP_032428.2 |
| AK149984.1 | 17 | 1883 | ||
| AK154307.1 | 5 | 378 | BAE32504.1 | |
| U14135.1 | 17 | 1864 | AAC52497.1 |
More Information
Set Membership: |
|

Genomic Map