Assay Details
Target Gene Details
Entrez Gene ID: | 69677 |
Gene Name: | interleukin 1 family, member 8 |
Gene Aliases: |
2310043N20Rik, Il36b |
Location: |
Chr.2:24153172-24160103 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il1f8 | NM_027163.4 | NP_081439.1 | ||
| AK009787.1 | BAB26505.1 | |||
| AY071842.1 | AAL67152.1 | |||
| BC119517.1 |
More Information
Set Membership: |
|

Genomic Map