Assay Details
Target Gene Details
Entrez Gene ID: | 16420 |
Gene Name: | integrin beta 6 |
Gene Aliases: |
2210409C20Rik, 4831415H04Rik |
Location: |
Chr.2:60598292-60722603 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb6 | NM_001159564.1 | NP_001153036.1 | ||
| NM_021359.3 | 1 | 174 | NP_067334.1 | |
| AF115376.1 | AAD17212.1 | |||
| AK008867.1 | 1 | 175 | ||
| AK019511.1 | BAB31769.1 | |||
| AK036439.1 | BAC29430.1 | |||
| AK085900.1 | ||||
| BC049185.1 | 1 | 142 | AAH49185.1 |
More Information
Set Membership: |
|

Genomic Map