Assay Details
Target Gene Details
Entrez Gene ID: | 16420 |
Gene Name: | integrin beta 6 |
Gene Aliases: |
2210409C20Rik, 4831415H04Rik |
Location: |
Chr.2:60598292-60722603 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itgb6 | NM_001159564.1 | 14 | 2113 | NP_001153036.1 |
| NM_021359.3 | 11 | 2007 | NP_067334.1 | |
| AF115376.1 | 14 | 1959 | AAD17212.1 | |
| AK036439.1 | 14 | 2114 | BAC29430.1 | |
| AK085900.1 | 14 | 2154 | ||
| BC049185.1 | 11 | 1976 | AAH49185.1 |
More Information
Set Membership: |
|

Genomic Map