Assay Details
Target Gene Details
Entrez Gene ID: | 227743 |
Gene Name: | mitogen-activated protein kinase associated protein 1 |
Gene Aliases: |
AI591529, D230039K05Rik, Sin1 |
Location: |
Chr.2:34406771-34624958 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapkap1 | NM_001290625.1 | NP_001277554.1 | ||
| NM_001290626.1 | NP_001277555.1 | |||
| NM_177345.4 | NP_796319.1 | |||
| AK027932.1 | BAC25671.1 | |||
| AK042154.1 | ||||
| AK052045.1 | BAC34838.1 | |||
| AK132263.1 | BAE21066.1 | |||
| BC027377.1 | AAH27377.1 | |||
| BC031579.1 | AAH31579.1 | |||
| BC043296.1 | AAH43296.1 | |||
| BC090644.1 | AAH90644.1 | |||
| BC096618.1 | AAH96618.1 |
More Information
Set Membership: |
|

Genomic Map