Assay Details
Target Gene Details
Entrez Gene ID: | 13589 |
Gene Name: | microtubule-associated protein, RP/EB family, member 1 |
Gene Aliases: |
5530600P05Rik, AI462499, AI504412, AW260097, BIM1p, D2Ertd459e, Eb1 |
Location: |
Chr.2:153741287-153773317 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapre1 | NM_007896.3 | NP_031922.1 | ||
| AK013626.1 | ||||
| AK146206.1 | ||||
| AK154245.1 | BAE32461.1 | |||
| BC052405.1 | AAH52405.1 | |||
| U51196.1 | AAA96320.1 |
More Information
Set Membership: |
|

Genomic Map