Assay Details
Target Gene Details
Entrez Gene ID: | 68975 |
Gene Name: | mediator complex subunit 27 |
Gene Aliases: |
1500015J03Rik, 2310042P07Rik, AA682045, Crsp8, D2Ertd434e |
Location: |
Chr.2:29346810-29524790 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med27 | NM_001290489.1 | 3 | 283 | NP_001277418.1 |
| NM_026896.5 | 4 | 557 | NP_081172.2 | |
| NM_029615.1 | 4 | 557 | NP_083891.1 | |
| AK005251.1 | 4 | 532 | BAB23906.2 | |
| AK009764.1 | 4 | 538 | BAB26488.2 | |
| AK147181.1 | 4 | 554 | BAE27743.1 | |
| BC016537.1 | 4 | 548 | AAH16537.1 | |
| CJ073494.1 | 4 | 355 |
More Information
Set Membership: |
|

Genomic Map