Assay Details
Target Gene Details
Entrez Gene ID: | 76467 |
Gene Name: | methionine sulfoxide reductase B2 |
Gene Aliases: |
2310050L06Rik, Mrsb, Msrb, Pilb |
Location: |
Chr.2:19371636-19394976 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msrb2 | NM_029619.2 | NP_083895.1 | ||
| AK009912.1 | ||||
| BC021619.1 | AAH21619.1 |
More Information
Set Membership: |
|

Genomic Map