Assay Details
Target Gene Details
Entrez Gene ID: | 17865 |
Gene Name: | myeloblastosis oncogene-like 2 |
Gene Aliases: |
B-Myb, Bmyb |
Location: |
Chr.2:163054333-163084688 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mybl2 | NM_008652.2 | NP_032678.1 | ||
| AK028497.1 | BAC25979.1 | |||
| BC050842.1 | AAH50842.1 | |||
| X70472.1 | CAA49898.1 |

Genomic Map