Assay Details
Target Gene Details
Entrez Gene ID: | 98932 |
Gene Name: | myosin, light polypeptide 9, regulatory |
Gene Aliases: |
AI327049, MLC20, Mylc2c, RLC-C |
Location: |
Chr.2:156775435-156781658 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myl9 | NM_172118.1 | NP_742116.1 | ||
| AK007972.1 | BAB25381.1 | |||
| AK020258.1 | BAB32043.1 | |||
| AK159880.1 | BAE35452.1 | |||
| BC049974.1 | ||||
| BC055439.1 | AAH55439.1 |
More Information
Set Membership: |
|

Genomic Map