Assay Details
Target Gene Details
Entrez Gene ID: | 78797 |
Gene Name: | NADPH dependent diflavin oxidoreductase 1 |
Gene Aliases: |
4930447P04Rik, NR1, Ndor |
Location: |
Chr.2:25244813-25255414 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndor1 | NM_001082476.2 | NP_001075945.1 | ||
| NM_001252541.1 | NP_001239470.1 | |||
| NM_001252542.1 | NP_001239471.1 | |||
| AK019617.1 | ||||
| AK143124.1 | ||||
| AK163207.1 | BAE37235.1 | |||
| AK169885.1 | BAE41435.1 | |||
| AK211498.1 | ||||
| BQ771207.1 |
More Information
Set Membership: |
|

Genomic Map