Assay Details
Target Gene Details
Entrez Gene ID: | 17932 |
Gene Name: | myelin transcription factor 1 |
Gene Aliases: |
Nzf2, Nzf2a, Nzf2b, Nztf2 |
Location: |
Chr.2:181763332-181827781 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myt1 | NM_001171615.1 | 20 | 3146 | NP_001165086.1 |
| NM_001171616.1 | 19 | 3256 | NP_001165087.1 | |
| NM_001171680.1 | 14 | 3336 | NP_001165151.1 | |
| NM_008665.4 | 20 | 3370 | NP_032691.2 | |
| AB082378.1 | 20 | 3357 | BAC16512.1 | |
| AB093267.2 | 19 | 2963 | BAC41451.2 | |
| AF004294.1 | 14 | 3337 | AAC53456.1 | |
| AK133358.1 | 12 | 1935 | ||
| AK135131.1 | 12 | 1532 | BAE22433.1 | |
| BC063252.1 | 20 | 3146 | AAH63252.1 |
More Information
Set Membership: |
|

Genomic Map