Assay Details
Target Gene Details
Entrez Gene ID: | 18286 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
AI848335, MMTEST29 |
Location: |
Chr.2:29889720-29931746 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Odf2 | NM_001113213.1 | NP_001106684.1 | ||
NM_001113214.1 | NP_001106685.1 | |||
NM_001177659.1 | NP_001171130.1 | |||
NM_001177661.1 | NP_001171132.1 | |||
NM_013615.3 | NP_038643.1 | |||
AF000968.1 | AAB65157.1 | |||
AF034105.1 | AAB87525.1 | |||
AK143726.1 | BAE25517.1 | |||
AK148523.1 | BAE28599.1 | |||
AK162550.1 | 1 | 360 | ||
AK165588.1 | ||||
AK168415.1 | BAE40330.1 | |||
BC057001.1 | AAH57001.1 | |||
DQ091767.1 | AAZ39867.1 | |||
DQ091770.1 | AAZ39870.1 |
