Assay Details
Target Gene Details
Entrez Gene ID: | 18286 |
Gene Name: | outer dense fiber of sperm tails 2 |
Gene Aliases: |
AI848335, MMTEST29 |
Location: |
Chr.2:29889720-29931746 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Odf2 | NM_001113213.1 | NP_001106684.1 | ||
| NM_001113214.1 | NP_001106685.1 | |||
| NM_001177659.1 | NP_001171130.1 | |||
| NM_001177661.1 | NP_001171132.1 | |||
| NM_013615.3 | NP_038643.1 | |||
| AF000968.1 | AAB65157.1 | |||
| AF034105.1 | AAB87525.1 | |||
| AK143726.1 | BAE25517.1 | |||
| AK148523.1 | BAE28599.1 | |||
| AK165588.1 | ||||
| AK168415.1 | BAE40330.1 | |||
| BC057001.1 | AAH57001.1 |

Genomic Map