Assay Details
Target Gene Details
Entrez Gene ID: | 241489 |
Gene Name: | phosphodiesterase 11A |
Gene Aliases: |
6330414F14Rik, A630086N24Rik, Gm350, PDE11A1 |
Location: |
Chr.2:75987026-76339145 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde11a | NM_001081033.1 | NP_001074502.1 | ||
| AK042383.1 | ||||
| BC158125.1 |
More Information
Set Membership: |
|

Genomic Map