Assay Details
Target Gene Details
Entrez Gene ID: | 18573 |
Gene Name: | phosphodiesterase 1A, calmodulin-dependent |
Gene Aliases: |
AI987702, AW125737 |
Location: |
Chr.2:79834453-80196518 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde1a | NM_001009978.1 | 2 | 144 | NP_001009978.1 |
| NM_001009979.1 | 2 | 144 | NP_001009979.1 | |
| NM_001159582.1 | 4 | 646 | NP_001153054.1 | |
| NM_016744.4 | 3 | 441 | NP_058024.2 | |
| AF023529.1 | 3 | 361 | AAB81952.1 | |
| AF159298.1 | 2 | 144 | AAF63857.1 | |
| AK014810.1 | 2 | 143 | ||
| AK043647.1 | 3 | 442 | BAC31606.1 | |
| AK050004.1 | 4 | 507 | ||
| AY845863.1 | 3 | 193 | ||
| AY845864.1 | 2 | 144 | AAW31879.1 | |
| BC090628.1 | 3 | 409 | AAH90628.1 | |
| CF741268.1 | 4 | 646 | ||
| U56649.1 | 4 | 377 | AAB03319.1 |
More Information
Set Membership: |
|

Genomic Map