Assay Details
Target Gene Details
Entrez Gene ID: | 18769 |
Gene Name: | protein kinase inhibitor, gamma |
Gene Aliases: |
PKIgamma |
Location: |
Chr.2:163658386-163726158 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pkig | NM_001039390.2 | NP_001034479.1 | ||
| NM_001039391.2 | NP_001034480.1 | |||
| NM_001164053.1 | NP_001157525.1 | |||
| NM_001164055.1 | NP_001157527.1 | |||
| NM_001277096.1 | NP_001264025.1 | |||
| NM_011106.2 | NP_035236.1 | |||
| AK135505.1 | ||||
| AK164716.1 | BAE37887.1 | |||
| AK168614.1 | BAE40480.1 | |||
| AK184424.1 | ||||
| AK186035.1 | ||||
| BC026550.1 | AAH26550.1 | |||
| BY240874.1 | ||||
| BY266193.1 | ||||
| BY283174.1 | ||||
| BY343638.1 | ||||
| CJ051558.1 | ||||
| U97170.1 | AAC09065.1 | |||
| W75470.1 |
More Information
Set Membership: |
|

Genomic Map